4-Nitrobenzamide ≥98%

189286-5G 189286-25G
220041-494EA 33.17 USD
220041-494 220041-493
4-Nitrobenzamide ≥98%
4-Nitrobenzamide
Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Melting Pt: 198 - 202 °C
Density: 1.59 g/cm³ (20 °C)
MDL Number: MFCD00007994
CAS Number: 619-80-7
EINECS: 210-613-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR