p-Toluenesulfonyl isocyanate ≥96%

Synonyms: Tosyl isocyanate, 4-Isocyanatosulphonyltoluene

189278-25G 189278-100G 189278-500G
220041-491EA 44.58 USD
220041-491 220041-490 220041-492
p-Toluenesulfonyl isocyanate ≥96%
p-Toluenesulphonyl isocyanate
Formula: C₈H₇NO₃S
MW: 197.21 g/mol
MDL Number: MFCD00002030
CAS Number: 4083-64-1
EINECS: 223-810-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR