2-Biphenylylmethanol ≥99%

Synonyms: 2-Biphenylmethanol, 2-Phenylbenzyl alcohol, [1,1-Biphenyl]-2-ylmethanol

188824-10G 188824-1G
220041-446EA 23.85 USD
220041-446 220041-447
2-Biphenylylmethanol ≥99%
2-Biphenylylmethanol
Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 41 - 42 °C
Density: 1.2 g/cm³ (20 °C)
MDL Number: MFCD00004620
CAS Number: 2928-43-0
EINECS: 220-892-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR