5-Amino-2-chloropyridine

Warning

Synonyms: 2-Chloro-5-pyridylamine, 6-Chloropyridin-3-amine, 2-Chloropyridin-5-amine

188778-5G 188778-1G
220041-443EA 59.98 USD
220041-443 220041-442
5-Amino-2-chloropyridine
5-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 83 °C
MDL Number: MFCD00006243
CAS Number: 5350-93-6
EINECS: 226-322-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR