3,5-Dichlorobenzyl alcohol ≥98%

188735-25G 188735-5G
220041-439EA 149.69 USD
220041-439 220041-440
3,5-Dichlorobenzyl alcohol ≥98%
3,5-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 82 °C
MDL Number: MFCD00004634
CAS Number: 60211-57-6
EINECS: 262-109-1
UN: 2250
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR