6-Nitro-o-toluonitrile ≥98%

Synonyms: 2-Methyl-6-nitrobenzonitrile, 6-Nitro-o-tolunitrile

188719-5G 188719-25G
220041-438EA 170.77 USD
220041-438 220041-437
6-Nitro-o-toluonitrile ≥98%
6-Nitro-o-toluonitrile
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD00007266
CAS Number: 1885-76-3
EINECS: 217-553-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR