2-(4-Hydroxyphenyl)ethanol ≥98%

Warning

Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol

188255-25G 188255-5G
220041-156EA 113.08 USD
220041-156 220041-157
2-(4-Hydroxyphenyl)ethanol ≥98%
2-(4-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 90…92 °C
MDL Number: MFCD00002902
CAS Number: 501-94-0
EINECS: 207-930-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR