1-Adamantaneethanol

Synonyms: 1-(2-Hydroxyethyl)adamantane, 1-(2-Hydroxyethyl)tricyclo[3.3.1.1(3,7)]decane, 2-(1-Adamantyl)ethanol

188115-1G 188115-5G
220041-141EA 57.11 USD
220041-141 220041-142
1-Adamantaneethanol
1-Adamantaneethanol
Formula: C₁₂H₂₀O
MW: 180.29 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 98.5 - 100 °C
MDL Number: MFCD00074756
CAS Number: 6240-11-5
EINECS: 228-353-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR