2-(3-Methoxyphenyl)ethanol ≥97%

Synonyms: 3-Methoxyphenethyl alcohol

187933-25G 187933-5G 187933-1G
220041-416EA 288.62 USD
220041-416 220041-417 220041-415
2-(3-Methoxyphenyl)ethanol ≥97%
2-(3-Methoxyphenyl)ethanol
Formula: C₉H₁₂O₂
MW: 152.19
MDL Number: MFCD00002893
CAS Number: 5020-41-7
EINECS: 225-705-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR