1-Bromo-3,5-dimethyladamantane

Synonyms: 1-Bromo-3,5-dimethyltricyclo[3.3.1.1(3,7)]decane, Memantine Related Compound D

187844-5G 187844-25G
220041-406EA 68.92 USD
220041-406 220041-405
1-Bromo-3,5-dimethyladamantane
1-Bromo-3,5-dimethyladamantane
Formula: C₁₂H₁₉Br
MW: 243.19 g/mol
MDL Number: MFCD00077197
CAS Number: 941-37-7
EINECS: 213-378-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR