2,6-Dichlorobenzoyl chloride ≥99%

Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

187518-50G 187518-10G
220041-393EA 274.62 USD
220041-393 220041-392
2,6-Dichlorobenzoyl chloride ≥99%
2,6-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000662
CAS Number: 4659-45-4
EINECS: 225-102-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR