3,4,5-Trichloroaniline ≥97%

187119-25G 187119-5G
220041-380EA 23.85 USD
220041-380 220041-381
3,4,5-Trichloroaniline ≥97%
3,4,5-Trichloroaniline
Formula: C₆H₄Cl₃N
MW: 196.46 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007769
CAS Number: 634-91-3
EINECS: 211-218-2
UN: 2018
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR