α,2-Dibromotoluene ≥98%

Danger

Synonyms: 2-Bromobenzyl bromide

187070-25G 187070-100G
220041-376EA 91.11 USD
220041-376 220041-375
α,2-Dibromotoluene ≥98%
α,2-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 34 °C
Density: 1.881 g/cm³ (20 °C)
Flash Pt: ≥ 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000173
CAS Number: 3433-80-5
EINECS: 222-334-8
UN: 2293
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR