α,3-Dibromotoluene ≥99%

Danger

Synonyms: 3-Bromobenzyl bromide, 1-Bromo-3-(bromomethyl)benzene

187062-5G 187062-25G
220041-374EA 53.95 USD
220041-374 220041-373
α,3-Dibromotoluene ≥99%
α,3-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 258 °C (1013 hPa)
Melting Pt: 41 - 44 °C
Density: 1.56 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000176
CAS Number: 823-78-9
EINECS: 212-519-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR