4-Bromobenzyl alcohol ≥99%

Synonyms: (4-Bromophenyl)methanol

187054-10G 187054-50G
220041-371EA 78.72 USD
220041-371 220041-372
4-Bromobenzyl alcohol ≥99%
4-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 77 - 78 °C
MDL Number: MFCD00004650
CAS Number: 873-75-6
EINECS: 212-851-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR