(S)-(+)-2-Amino-3-methyl-1-butanol ≥96%

Synonyms: L(+)-2-Amino-3-methyl-1-butanol, (S)-2-Amino-3-methylbutan-1-ol, (S)-2-Amino-3-methyl-butan-1-ol, L-Valinol, L(+)-Valinol

186708-5G 186708-25G 186708-1G
220041-361EA 91.11 USD
220041-361 220041-360 220041-359
(S)-(+)-2-Amino-3-methyl-1-butanol ≥96%
(S)-(+)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 31…32 °C
Density: 0.92 g/cm³ (20 °C)
MDL Number: MFCD00064296
CAS Number: 2026-48-4
EINECS: 217-975-5

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR