18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)

Warning

186651-25G 186651-5G
220041-354EA 286.46 USD
220041-354 220041-355
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.237 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5
Merck Index: 13,02630

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR