N,N-Dimethyl-p-phenylenediammonium sulfate ≥98%

Danger

Synonyms: N,N-Dimethyl-1,4-phenylenediammonium sulphate, N,N-Dimethylbenzene-1,4-diammonium sulphate

186384-100G 186384-25G
220040-979EA 269.23 USD
220040-979 220040-980
N,N-Dimethyl-p-phenylenediammonium sulfate ≥98%
N,N-Dimethyl-p-phenylenediammoniumsulphate
Formula: C₈H₁₄N₂O₄S
MW: 234.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012992
CAS Number: 536-47-0
EINECS: 208-636-2
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR