5-Aminoisophthalic acid ≥94%

Warning

186279-100G 186279-25G
220040-228EA 83.78 USD
220040-228 220040-229
5-Aminoisophthalic acid ≥94%
5-Aminoisophthalic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 515 °C (1013 hPa)
Melting Pt: 150 - 156 °C
MDL Number: MFCD00002522
CAS Number: 99-31-0
EINECS: 202-748-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR