Dimethyl adipate ≥99%

Synonyms: Adipic acid dimethyl ester, 1,6-Hexanedioic acid dimethyl ester

186252-100G 186252-500G
220040-968EA 56.88 USD
220040-968 220040-969
Dimethyl adipate ≥99%
Dimethyl adipate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: ∼229 °C (1013 hPa)
Melting Pt: 10.3 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008469
CAS Number: 627-93-0
EINECS: 211-020-6
Merck Index: 13,00163

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR