Dimethyl sulfoxide-[D₆]

Synonyms: Dimethyl sulphoxide-[²H₆], Hexadeuterodimethyl sulphoxide, DMSO-D6, DMSO-d6, Dimethyl sulphoxide-d6

185965-50G 185965-25G 185965-10G
220041-210EA 70.77 USD
220041-210 220041-209 220041-208
Dimethyl sulfoxide-[D₆]
Dimethyl sulphoxide-[D₆]
Formula: D₃CS(O)CD₃
MW: 84.09 g/mol
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 82 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR