2-Chloro-2-methylpropane-[D9]

Synonyms: tert-Butyl chloride-D9

185949-1G 185949-5G
220041-138EA 236.31 USD
220041-138 220041-139
2-Chloro-2-methylpropane-[D9]
2-Chloro-2-methylpropane-[D9]
Formula: D₃CCCl(CD₃)CD₃
MW: 101.5 g/mol
CAS Number: 918-20-7
EINECS: 213-041-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR