Furfuryl alcohol

Danger

Synonyms: Furfurol

185930-250G 185930-50G 185930-1KG
220041-136EA 38.55 USD
220041-136 220041-137 220041-135
Furfuryl alcohol
Furfuryl alcohol
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: -31 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003252
CAS Number: 98-00-0
EINECS: 202-626-1
UN: 2874
ADR: 6.1,III
Merck Index: 13,04326

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR