Furan

Danger

185922-500ML 185922-100ML 185922-5ML
220041-133EA 76.89 USD
220041-133 220041-132 220041-134
Furan
Furan
Formula: C₄H₄O
MW: 68.08 g/mol
Boiling Pt: 31.4 °C (1013 hPa)
Melting Pt: -86 °C
Density: 0.9371 g/cm³ (20 °C)
Flash Pt: -50 °C
MDL Number: MFCD00003222
CAS Number: 110-00-9
EINECS: 203-727-3
UN: 2389
ADR: 3,I
Merck Index: 13,04317

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR