N,N-Diethylaniline ≥99%

Danger

185868-50ML 185868-1L 185868-250ML
220041-127EA 17.66 USD
220041-127 220041-125 220041-126
N,N-Diethylaniline ≥99%
N,N-Diethylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 215.5 °C (1013 hPa)
Melting Pt: –38.8 °C
Density: 0.9348 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00009042
CAS Number: 91-66-7
EINECS: 202-088-8
UN: 2432
ADR: 6.1,III
Merck Index: 13,03140

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR