Cyanuric acid

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

185809-100G 185809-25G
220041-117EA 61.6 USD
220041-117 220041-118
Cyanuric acid
Cyanuric acid
Formula: C₃H₃N₃O₃
MW: 129.08 g/mol
Density: 2.5 g/cm³ (20 °C)
MDL Number: MFCD00082990
CAS Number: 108-80-5
EINECS: 203-618-0
Merck Index: 13,02728

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR