1-Chloro-2-nitrobenzene ≥99%

Danger

Synonyms: 1-Chloro-2-nitrobenzene, 2-Chloronitrobenzene

185760-1KG 185760-250G
220041-108EA 82.71 USD
220041-108 220041-109
1-Chloro-2-nitrobenzene ≥99%
1-Chloro-2-nitrobenzene
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 33 °C
Density: 1.368 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007061
CAS Number: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: 6.1,II
Merck Index: 13,02170

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR