1-Chloronaphthalene, Technical Grade

Warning

Synonyms: Triisodecyl phosphite, α-Chloronaphthalene

185752-500G 185752-100G
220041-107EA 156.15 USD
220041-107 220041-106
1-Chloronaphthalene, Technical Grade
1-Chloronaphthalene
Formula: C₁₀H₇Cl
MW: 162.62 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: -2.5 °C
Density: 1.1938 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00003874
CAS Number: 90-13-1
EINECS: 201-967-3
UN: 2810
ADR: 6.1,III
Merck Index: 13,02168

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR