5-Fluoro-1-indanone

Synonyms: 5-Fluoro-2,3-dihydro-1H-inden-1-one

185663-5G 185663-1G
220041-034EA 472.77 USD
220041-034 220041-033
5-Fluoro-1-indanone
5-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.15 g/mol
Boiling Pt: 113…114 °C (10 mmHg)
Melting Pt: 35…37 °C
Density: 1.35 g/cm³ (20 °C)
MDL Number: MFCD00041031
CAS Number: 700-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR