Propiononitrile ≥99%

Synonyms: Propionitrile

185590-100ML 185590-5ML 185590-500ML
220040-580EA 62.35 USD
220040-580 220040-582 220040-581
Propiononitrile ≥99%
Propiononitrile
Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 97.2 °C (1013 hPa)
Melting Pt: -93 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00001948
CAS Number: 107-12-0
EINECS: 203-464-4
UN: 2404
ADR: 3,II
Merck Index: 12,08012

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR