Benzyl(dimethyl)amine ≥99%

Danger

Synonyms: DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA

185582-500ML 185582-100ML
220040-579EA 94.55 USD
220040-579 220040-578
Benzyl(dimethyl)amine ≥99%
Benzyl(dimethyl)amine
Formula: C₆H₅CH₂N(CH₃)₂
MW: 135.21 g/mol
Boiling Pt: 183 °C (1013 hPa)
Melting Pt: -75 °C
Density: 0.915 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008329
CAS Number: 103-83-3
EINECS: 203-149-1
UN: 2619
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90635bbe74e3372ed4bfbd8d750b25afd50f77a5&vid=7044105a7e7fbce7e00f1d15d7a4e017a1b9d418&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR