N-Benzylaniline ≥99%

Synonyms: N-Phenylbenzylamine, ((Phenylmethyl)amino)benzene

185493-100G 185493-25G
220040-286EA 202.46 USD
220040-286 220040-287
N-Benzylaniline ≥99%
N-Benzylaniline
Formula: C₆H₅CH₂NHC₆H₅
MW: 183.25 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 33…36 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00003018
CAS Number: 103-32-2
EINECS: 203-100-4
Merck Index: 12,01161
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cee0f9ae18e00188e4a272cc12dbca2071f87d96&vid=22e30683618c7daab6a3a3013140064b6131ccea&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR