o-Toluidine ≥99%

Danger

Synonyms: 2-Toluidine, 2-Aminotoluene, 2-Methylaniline

185426-5G 185426-100G 185426-500G
220040-573EA 46.2 USD
220040-573 220040-571 220040-572
o-Toluidine ≥99%
o-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: < –15 °C
Density: 0.998 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007730
CAS Number: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1,II
Merck Index: 12,09674

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR