(±)-Tetrahydrofurfuryl alcohol

Warning

Synonyms: (±)-Tetrahydro-2-furfuryl alcohol, (Tetrahydrofuran-2-yl)methanol, (±)-Tetrahydro-2-furyl methanol, (±)-THFA

185396-5G 185396-250G 185396-1KG
220040-570EA 29.72 USD
220040-570 220040-569 220040-568
(±)-Tetrahydrofurfuryl alcohol
(±)-Tetrahydrofurfuryl alcohol
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: < –80 °C
Density: 1.0543 g/cm³ (20 °C)
Flash Pt: 75 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005372
CAS Number: 97-99-4
EINECS: 202-625-6
Merck Index: 12,09353

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR