Terephthalic acid ≥98%

185361-100G 185361-5G 185361-500G
220041-028EA 49.95 USD
220041-028 220041-030 220041-029
Terephthalic acid ≥98%
Terephthalic acid
Formula: C₈H₆O₄
Boiling Pt: 392 °C (1013 hPa)
Density: 1.472 g/cm³ (20 °C)
Flash Pt: 260 °C
Storage Temperature: Ambient
MDL Number: MFCD00002558
CAS Number: 100-21-0
EINECS: 202-830-0
Merck Index: 12,09306

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR