Phenyl isocyanate ≥98%

Danger

185353-5G 185353-500G 185353-100G
220041-027EA 34.68 USD
220041-027 220041-026 220041-025
Phenyl isocyanate ≥98%
Phenyl isocyanate
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: -33 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00001994
CAS Number: 103-71-9
EINECS: 203-137-6
UN: 2487
ADR: 6.1,I
Merck Index: 12,07449

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR