3-Nitrobenzoic acid ≥99%

Warning

Synonyms: m-Nitrobenzoic acid

185329-5G 185329-500G 185329-100G
220041-024EA 39.85 USD
220041-024 220041-023 220041-022
3-Nitrobenzoic acid ≥99%
3-Nitrobenzoic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 341 °C (1013 hPa)
Melting Pt: 139…142 °C
Density: 1.494 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007251
CAS Number: 121-92-6
EINECS: 204-508-5
Merck Index: 12,06686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR