4-Nitroaniline ≥99%

Danger

Synonyms: p-Nitroaniline, pNA

185310-500G 185310-100G
220041-021EA 86.15 USD
220041-021 220041-020
4-Nitroaniline ≥99%
4-Nitroaniline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 148 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00007858
CAS Number: 100-01-6
EINECS: 202-810-1
UN: 1661
ADR: 6.1,II
Merck Index: 12,06681

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR