Allyl acetate ≥99%

185248-100ML 185248-500ML
220040-953EA 78.72 USD
220040-953 220041-015
Allyl acetate ≥99%
Allyl acetate
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 103 °C (1013 hPa)
Melting Pt: –96 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008721
CAS Number: 591-87-7
EINECS: 209-734-8
UN: 2333
ADR: 3,II
Merck Index: 12,00293

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR