Dimethyl terephthalate ≥99%

185124-3KG 185124-500G
220040-513EA 152.92 USD
220040-513 220040-514
Dimethyl terephthalate ≥99%
Dimethyl terephthalate
Formula: C₁₀H₁₀O₄
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: 139 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00008440
CAS Number: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR