2-(Hydroxymethyl)benzyl alcohol ≥97%

Synonyms: o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol

184829-1G 184829-10G
220040-937EA 80.68 USD
220040-937 220040-936
2-(Hydroxymethyl)benzyl alcohol ≥97%
2-(Hydroxymethyl)benzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 62…64 °C
MDL Number: MFCD00004626
CAS Number: 612-14-6
EINECS: 210-293-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR