4-Phenyl-1-butanol ≥99%

Synonyms: 4-Phenylbutan-1-ol

184756-10G 184756-50G
220040-930EA 140 USD
220040-930 220040-931
4-Phenyl-1-butanol ≥99%
4-Phenyl-1-butanol
Formula: (C₆H₅)(CH₂)₃CH₂OH
MW: 150.22 g/mol
Boiling Pt: 140 °C (1013 hPa)
Density: 0.975 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS Number: 3360-41-6
EINECS: 222-128-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR