(Chloromethyl)cyclopropane ≥97%

Synonyms: Cyclopropyl methyl chloride

184667-50G 184667-10G
220040-924EA 691.38 USD
220040-924 220040-923
(Chloromethyl)cyclopropane ≥97%
(Chloromethyl)cyclopropane
Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 87 °C (1013 hPa)
Melting Pt: -91 °C
Density: 0.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001307
CAS Number: 5911-08-0
EINECS: 227-632-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR