Petroleum spirit, 40…60 °C ACS, Sigma-Aldrich®

Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

184519-500ML 184519-4L 184519-1L
220040-913EA 17.23 USD
220040-913 220040-912 220040-224
Petroleum spirit, 40…60 °C ACS, Sigma-Aldrich®
Petroleum spirit
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 1268
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR