2-Bromobenzyl alcohol ≥99%

Synonyms: (2-Bromophenyl)methanol

184276-50G 184276-10G
220040-221EA 132.46 USD
220040-221 220040-220
2-Bromobenzyl alcohol ≥99%
2-Bromobenzyl alcohol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 80 - 82 °C
MDL Number: MFCD00004600
CAS Number: 18982-54-2
EINECS: 242-719-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR