11-Bromo-1-undecanol ≥98%

Synonyms: 11-Bromoundecan-1-ol, 11-Bromoundecanol

184136-10G 184136-50G
220040-205EA 68.71 USD
220040-205 220040-206
11-Bromo-1-undecanol ≥98%
11-Bromo-1-undecanol
Formula: HO(CH₂)₁₀CH₂Br
MW: 251.21 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00004752
CAS Number: 1611-56-9
EINECS: 216-554-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR