2'-Chloroacetophenone ≥97%

Warning

Synonyms: 1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone

183709-100G 183709-25G
220040-884EA 70.11 USD
220040-884 220040-885
2'-Chloroacetophenone ≥97%
2'-Chloroacetophenone
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 227 °C (1013 hPa)
Density: 1.189 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000560
CAS Number: 2142-68-9
EINECS: 218-397-6
UN: 3416
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR