2-Iodobenzyl alcohol ≥99%

183636-10G 183636-50G
220040-872EA 87.02 USD
220040-872 220040-873
2-Iodobenzyl alcohol ≥99%
2-Iodobenzyl alcohol
Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 145 °C (10 mmHg)
Melting Pt: 91 °C
MDL Number: MFCD00004610
CAS Number: 5159-41-1
EINECS: 225-933-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR