5-Methoxy-1-indanone

183539-5G 183539-10G
220040-867EA 124.92 USD
220040-867 220040-866
5-Methoxy-1-indanone
5-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 145 - 150 °C (0.2 -rr)
Melting Pt: 107 - 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003789
CAS Number: 5111-70-6
EINECS: 225-838-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR