2-(4-Chlorophenyl)ethanol ≥99%

Synonyms: 4-Chlorophenethyl alcohol

183423-10G 183423-50G
220040-850EA 107.69 USD
220040-850 220040-851
2-(4-Chlorophenyl)ethanol ≥99%
2-(4-Chlorophenyl)ethanol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 259 °C (1013 hPa)
Density: 1.189 g/cm³ (25 °C)
MDL Number: MFCD00002899
CAS Number: 1875-88-3
EINECS: 217-506-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR